Kalsiumhydroksid ≥98.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Slaked lime, Calcium dihydroxide

10304KAEA 112025 NOK
10304KA
Kalsiumhydroksid ≥98.0%, AnalaR NORMAPUR® analytical reagent
Kalsiumhydroksid
Formel: Ca(OH)₂
MW: 74.09 g/mol
Smeltepunkt: 550
Tetthet: 2.24 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010901
CAS-nummer: 1305-62-0
EINECS: 215-137-3
FN: 3262
ADR: 8 III
Merck Indeks: 13,01675

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98.0 %
Insoluble in hydrochloric acid Max. 0.02 %
Cl (Chloride) Max. 0.1 %
SO4 (Sulphate) Max. 0.01 %
Total N (Nitrogen) Max. 0.005 %
As (Arsenic) Max. 0.0001 %
Ba (Barium) Max. 0.005 %
Cu (Copper) 0.001 %
Fe (Iron) Max. 0.005 %
K (Potassium) Max. 0.05 %
Mg (Magnesium) Max. 0.05 %
Na (Sodium) Max. 0.05 %
Pb (Lead) Max. 0.001 %
Sr (Strontium) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR