Lantan(III)klorid heptahydrat ≥98%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals

103433QEA 1325 NOK
103433Q
Lantan(III)klorid heptahydrat ≥98%, AnalaR NORMAPUR® analytical reagent
Lantan(III)klorid heptahydrat
Formel: LaCl₃·7H₂O
MW: 371.37 g/mol
Kokepunkt: < 860 °C (1013 hPa)
Smeltepunkt: 96
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149756
CAS-nummer: 10025-84-0
EINECS: 233-237-5

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98 %
Identification Passes test
Insolubility in water Max. 0.1 %
Ca (Calcium) Max. 10 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 1 ppm
Pb (Lead) Max. 2 ppm
Zn (Zinc) Max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR