Natriumazid ≥99.0%, AnalaR NORMAPUR® analytisk reagens

Leverandør: VWR Chemicals
Danger

103692KEA 1025 NOK
103692K 103694M 10369AS
Natriumazid ≥99.0%, AnalaR NORMAPUR® analytisk reagens
Sodium azide
Formel: NaN₃
Smeltepunkt: 275 °C
Tetthet: 1,85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003536
CAS-nummer: 26628-22-8
EINECS: 247-852-1
FN: 1687
ADR: 6.1,II
Merck Indeks: 13,08653

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Appearance White crystalline powder
Identification Passes test
Free alkalinity Max. 0.05 meq/g
Insolubility in water Max. 0.015 %
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Cr (Chromium) Max. 20 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR