Certipur® ICP Multi Element Standard Solution IV
Leverandør: Merck
1044980100
1044980100.EA 0 NOK
1044980100.
Certipur® ICP Multi Element Standard Solution IV
Standarder Kationstandarder Multi-Element Standards
Sertifiseringer: This certified reference material (CRM) is produced in accordance with ISO 17034 and characterised in accordance with ISO/IEC 17025.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Hardhetsklasse beskrivelse Corrosive
UN No. 2031
GHS-signal Danger
Lagringstemp 15...25 °C