Jod ≥99.9985% (metals basis), krystaller, Puratronic®, resublimert

Leverandør: Thermo Fisher Scientific
Warning

10619.14 10619.22 10619.36 10619.88
10619.14EA 1975 NOK
10619.14 10619.22 10619.36 10619.88
Jod ≥99.9985% (metals basis), krystaller, Puratronic®, resublimert
Jod
Formel: I₂
Kokepunkt: 184,3 °C (1013 hPa)
Smeltepunkt: 183…186 °C (subl.)
Tetthet: 4,930
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011355
CAS-nummer: 7553-56-2
EINECS: 231-442-4
FN: 3495
ADR: 8,III
Merck Indeks: 13,05036

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Grey
Form Crystalline
Total Metal Impurities ≤0.0015% (15 ppm)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR