Zink ≥99.999% (metals basis), fragmenter 1-5 mm (0.04-0.2 in)

Leverandør: Thermo Fisher Scientific

10759.18 10759.30 10759.A1
10759.18EA 1475 NOK
10759.18 10759.30 10759.A1
Zink ≥99.999% (metals basis), fragmenter 1-5 mm (0.04-0.2 in)
Zinc

Purity: 99.999% CAS No.: 7440-66-6 Odour: Odourless MDL No.: MFCD00011291

Formel: Zn
Kokepunkt: 908 °C (1013 hPa)
Smeltepunkt: 420 °C
Tetthet: 7,14 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011291
CAS-nummer: 7440-66-6
EINECS: 231-175-3
Merck Indeks: 12,10255

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR