Svovel ≥99.5%, pulver, utfelt

Leverandør: Thermo Fisher Scientific
Danger

10785.36 10785.A3
10785.36EA 850 NOK
10785.36 10785.A3
Svovel ≥99.5%, pulver, utfelt
Svovel

Purity: 99.5% CAS No.: 7704-34-9 Odour: Weak UN No.: 1350 MDL No.: MFCD00085316

Formel: S
Kokepunkt: 445 °C (1013 hPa)
Smeltepunkt: 113…119 °C
Tetthet: 2,36 g/cm³ (20 °C)
Flash punkt: 168 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085316
CAS-nummer: 7704-34-9
EINECS: 231-722-6
FN: 1350
ADR: 4.1,III
Merck Indeks: 13,09059

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR