Palladium ≥99.995% (metals basis), pulver, Premion® , -22 mesh

Leverandør: Thermo Fisher Scientific
Warning

10788.03 10788.06 10788.93
10788.03EA 5175 NOK
10788.03 10788.06 10788.93
Palladium ≥99.995% (metals basis), pulver, Premion® , -22 mesh
Palladium

Purity: 99.995% CAS No.: 7440-05-3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167

Formel: Pd
Tetthet: 12,02 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011167
CAS-nummer: 7440-05-3
EINECS: 231-115-6
FN: 3089
ADR: 4.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR