Calcium Levulinate, Reference Standard (AS)

Leverandør: USP

1086935
1086935.EA 2225 NOK
1086935.
Calcium Levulinate, Reference Standard (AS)
Standarder Organiske standarder

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Sertifiseringer: US Pharmacopea Reference Standard

Bestill nå

SPESIFIKASJONER

Beskrivelse Calcium Levulinate
Environmentally Preferable
CAS-nr 5743-49-7
Formel C₁₀H₁₄CaO₆ · ₂H₂O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR