Eddiksyre ≥99.9985% (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

10994.AE 10994.AP 10994.AC
10994.AEEA 2450 NOK
10994.AE 10994.AP 10994.AC
Eddiksyre ≥99.9985% (metals basis)
Eddiksyre glasial (iseddik)

Purity: 99.9985% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Formel: H₃CCOOH
Kokepunkt: 118,1 °C
Tetthet: 1,049
Flash punkt: 40 °C (104 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
FN: 2789
ADR: 8,II
REACH: 01-2119475328-30
Merck Indeks: 14,55
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR