Palladium (II) oksid, vannfri ≥99.9% (metals basis) Pd ≥ 85%

Leverandør: Thermo Fisher Scientific
Warning

11040.03 11040.06
11040.03EA 4500 NOK
11040.03 11040.06
Palladium (II) oksid, vannfri ≥99.9% (metals basis) Pd ≥ 85%
Palladium (II) oksid

Purity: 99.9% CAS No.: 1314-08-5 Molecular Formula: PdO Formula Weight: 122.40 Physical Form: -22 Mesh Powder Density: 8.70 Appearance: Brown Odour: Odourless Melting Point: 870° Solubility: Insoluble in water, acids. Slightly soluble in aqua regia Merck Reference: 14,6993 MDL No.: MFCD00011172


Catalytic reduction, in organic synthesis

Formel: OPd
Smeltepunkt: 870 °C
Tetthet: 8,70
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011172
CAS-nummer: 1314-08-5
EINECS: 215-218-3
Merck Indeks: 14,6993

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR