Platinium(IV) oxide hydrate 99.9% (71 - 75% Pt) (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

11049.01 11049.03 11049.06 11049.14
11049.01EA 1025 NOK
11049.01 11049.03 11049.06 11049.14
Platinium(IV) oxide hydrate 99.9% (71 - 75% Pt) (metals basis)
Platinium(IV) oxide hydrate

Purity: 99.9% CAS No.: 52785-06-5 Molecular Formula: O2Pt.xH2O Formula Weight: 227.09(anhy) Physical Form: Powder Appearance: Dark brown Merck Reference: 14,7527 UN No.: 1479 MDL No.: MFCD00066964

Formel: O₂Pt·H₂O
Smeltepunkt: 450 °C
Tetthet: 10,2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066964
CAS-nummer: 52785-06-5
EINECS: 215-223-0
FN: 1479
ADR: 5.1,II
Merck Indeks: 14,7527

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR