Partikkelskivefiltre i 2000-serien for masker i 6000-/7000-/7500-serien
Leverandør: 3M
2125 2128 2135 2138 2128UK
111-0069EA 2625 NOK
111-0069 111-0070 111-0055 111-0056 111-2181
Partikkelskivefiltre i 2000-serien for masker i 6000-/7000-/7500-serien
Respiratorer Flerbruksmasker Filter for vernemasker
These mask filters are light, comfortable and provide reliable protection from particulates. When using with 3M™ 6000 series masks, a 3M™ 502 adapter is required.
  • Low breathing resistance
  • Allow a very wide field of vision
  • Bayonet fitting system ensures precise and secure locking
EN 143
Sertifiseringer: EN 143