Engangs partikkelfiltrerende åndedrettsvern, FFP3, 8833
Leverandør: 3M
8833
111-0451EA 2100 NOK
111-0451
Engangs partikkelfiltrerende åndedrettsvern, FFP3, 8833
Respiratorer Engangsmasker Støvmasker
The robust shell and cup design of the mask conforms well to most face shapes and sizes, and maintains its shape during use. The excellent fit is provided by the convex shape, the nose clip, and the twin-strap design.
  • Cool Flow™ valve reduces heat build-up to offer comfortable protection, particularly in hot and humid conditions
  • Soft inner face-seal ring provides secure face fit and increases wearer comfort
  • Reliable, effective protection against fine particulates
  • Comfortable, lightweight off-the-face design
  • Red colour coded straps denoting FFP3 protection