VWR®, Cleanroom Disposable Goggles
Leverandør: VWR Collection
111-0615EA 840 NOK
111-0615
VWR®, Cleanroom Disposable Goggles
Vernebriller /Goggles
Sterile cleanroom goggles with a lightweight frame, indirect ventilation, and a non linting, latex-free headband.
  • Toughened PC lens with anti-fog and anti-scratch coating
  • Lightweight PVC frame with silicone rubber strap provides comfort
  • Ethylene oxide (EtO) sterilised and fully validated to ISO 11135-1-2007
Recommended for ISO Class 4 and EU GMP Grade A/B critical environments.
Sertifiseringer: EN 166:2001, Optical Class 1.
Forpakning: In a Nextech certified ISO Class 4 for cleanroom. One piece per Tyvek™ PE bag.