Halvmasker for partikkelfiltrering, til engangsbruk, FFP1/FFP2/FFP3, Aura™ 9300+
Leverandør: 3M
9310PLUS 9322PLUS 9312PLUS 9320PLUS 9330PLUS 9332PLUS GT500073439 9320PLUSSPBULK 9330PLUSBV 9332PLUSBV
111-0713EA 585 NOK
111-0713 111-0716 111-0714 111-0715 111-0717 111-0684 111-2201 111-2621 111-2534 111-2533
Halvmasker for partikkelfiltrering, til engangsbruk, FFP1/FFP2/FFP3, Aura™ 9300+
Respiratorer Engangsmasker Støvmasker
3M™ Aura™ Particulate Respirators 9300+ Series has an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels, and with valved and unvalved models, these respirators feature an innovative chin tab for easy adjustments and an embossed, sculpted upper panel to help reduce eyewear fogging.
  • Comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments
  • FFP1, FFP2, and FFP3 protection offered, allowing users to talk