Disposable particle filtering respirators, FFP2, Aura™ 9400+
Leverandør: 3M
9422PLUS
111-0718EA 835 NOK
111-0718
Disposable particle filtering respirators, FFP2, Aura™ 9400+
Respiratorer Engangsmasker Støvmasker
Three-panel design fits a wide range of face shapes and sizes to accommodate individual facial movements.
  • Staple-free: Helps to minimise contamination risks
  • Metal detectable: Embedded metal nose clip to help ensure production safety
  • Concealed adjustable nose clip: Kept securely in place within the layers of the respirator
  • Cool Flow™ valve effectively removes heat build-up, removes exhaled air and minimises the risk of fogging eyewear
  • Flat-fold; easy to store when not in use
Colour: Blue
Sertifiseringer: EN 149:2001+A1:2009
NB: *One size fits all