Goggles, Muria
Leverandør: DELTA PLUS
MURIA1VD
111-0905EA 91 NOK
111-0905
Goggles, Muria
Vernebriller /Goggles
These goggles have PVC transparent frames and PC lenses for protection against impacts caused by medium energy (impact 120 m/s), during grinding work.
  • Woven elastic band
  • Direct ventilation
  • Soft PVC nose bridge
Weight: 60 g
Sertifiseringer: Certified with EN 166 1 BT / BT.