Banded earplugs, E-A-Rcap™
Leverandør: 3M
EC01000
111-0912EA 7575 NOK
111-0912
Banded earplugs, E-A-Rcap™
Ørepropper Ørepropper med bøyle
Banded earplugs are a practical option and easy to use.
  • Extremely lightweight, virtually no feeling of pressure
  • Plugs do not push into your ear canal; prevents irritation
  • Ideal for frequent insertion and removal
They can be worn around the neck when not needed, making them ideal for temporary use.
SNR: 23 dB
Sertifiseringer: EN 352-2