Safety helmet, Granite Wind
Leverandør: DELTA PLUS
GRAWIJAFL
111-0928EA 900 NOK
111-0928
Safety helmet, Granite Wind
Hjelmer Sikkerhethjelm
Ventilated construction helmet made of ABS and designed like a climbing helmet. Fluorescent yellow colour provides a good visibility.
  • Without helmet shield for better vertical view
  • Textile interior and Thermoform lining
  • Adjustable rotor system for easy tightening
  • Chin strap with three fastening points
Size: 530 to 630 mm
Weight: 355 g
EN 397
Sertifiseringer: EN 397