VWR® Dublin, Safety Spectacles
Leverandør: VWR Collection
111-0929EA 120 NOK
111-0929
VWR® Dublin, Safety Spectacles
Vernebriller
Lightweight spectacles with clear polycarbonate lens.
  • Anti-scratch/anti-fog coating
  • Moulded brow guard
  • Soft nose pad
  • Soft rubber temple pads
Weight: 25,5 g
EN 166, EN 170; 1 FT
Sertifiseringer: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)