Goggles, Blast
Leverandør: Bollé Safety
BLAPSI
111-0983EA 0 NOK
111-0983
Goggles, Blast
Vernebriller /Goggles
These goggles offer an outstanding field of vision, fit over prescription spectacles and can be worn with respiratory half masks. They offer unbeatable comfort and essential modular design. In combination with the accessory visor, this model can also be used as a face shield.
  • Liquid overflow chute
  • Ball-pivoting, adjustable strap
  • Available with or without foam, ventilated or sealed, with PC or acetate lens
Vekt: 113 g
Sertifiseringer: EN 166