Harnesses, Revolution Premium R7
Leverandør: HONEYWELL SAFETY
1014807/OFFS 1014808/OFFS 1014809/OFFS
111-1417EA 12950 NOK
111-1417 111-1418 111-1419
Harnesses, Revolution Premium R7
Fallvern Fallsele
These 4-point harnesses are equipped with Dualtech webbing, rotating work positioning belts, automatic buckles and Ergo Armor™ back shields. They are ideal for wind energy applications, building and construction.
  • Pivot Link system for better ergonomic fit when bending or squatting
  • Dualtech webbing (polyamide/polyester) with shape-retention memory
  • Padded work positioning belt for more comfort
  • Easy and quick adjustment
Sertifiseringer: EN 361, EN 358.