Disposable particle filtering respirators, FFP1/FFP2/FFP3, folded, 2000 series
Leverandør: HONEYWELL SAFETY
1031594-V2 1031590 1031591
111-1544EA 1400 NOK
111-1544 111-1540 111-1541
Disposable particle filtering respirators, FFP1/FFP2/FFP3, folded, 2000 series
Respiratorer Engangsmasker Støvmasker
Masks with elasticated fasteners and low-profile shape guarantee a perfect vision. Available with or without valve.
  • Wraparound shape with large filter surface and increased comfort
  • No staples, no risk of irritation
  • Coloured nose-bridge ensures easy adjustment and instant identification of the level of protection
  • Willtech seal: hypoallergenic, anti-perspirant, better comfort
  • Individual reusable bag protects the mask before use and during breaks
  • PVC, silicone and latex-free
EN 149 + A1*
Sertifiseringer: EN 149 + A1*