Autoclavable cleanroom goggles, uvex ultrasonic CR
Leverandør: UVEX
9302.500
111-1596EA 1375 NOK
111-1596
Autoclavable cleanroom goggles, uvex ultrasonic CR
Vernebriller /Goggles
These goggles provide wearers with excellent comfort, functionality and an innovative design. Suitable for use over prescription spectacles.
  • Ultra-light autoclavable goggles with anti-fog effect on inner lens surface
  • Autoclavable at least 10 times at 121 °C for 20 min
  • Non allergenic material - free of harmful substances
  • Panoramic lens, duo component technology and adjustable silicone headband
  • Indirect ventilation ensures a pleasant and comfortable interior environment
  • Easy lens replacement
Designed for use in cleanrooms and in food processing, medicalMore Product Information