Cleanroom safety spectacles, autoclavable, super fit CR
Leverandør: UVEX
9178.500
111-1598EA 250 NOK
111-1598
Cleanroom safety spectacles, autoclavable, super fit CR
Vernebriller
These single lens safety glasses provide eye protection, comfort and functionality with an innovative design. They are suitable for use in combination with other PPE (e.g. face masks).
  • Very light autoclavable protective spectacles with anti-fog effect on inner lens surface
  • Autoclavable at least 10 times at 121 °C for 20 min
  • Non allergenic material - free of harmful substances
  • Ultra-thin wrap-around lens with extremely good coverage of brow and cheek area
  • XST side arms for increased comfort and pressure-free fit
Ideal for use in food processing,More Product Information