Reusable earplugs, E-A-R™ Ultrafit™ X
Leverandør: 3M
UF01014
111-1629EA 3950 NOK
111-1629
Reusable earplugs, E-A-R™ Ultrafit™ X
Ørepropper Flergangsørepropper
Corded earplugs made from a patented polymer formulation, the comfortable triple flanged tip of these earplugs provides an excellent attenuation of 35 dB SNR, one of the highest amongst pre-moulded earplugs. The earplugs are already pre-formed to ensure optimum fit.
  • Triple-flange feature of the earplugs ensures a perfect fit: No size adjustment required
  • Fit most ear canals comfortably and provide a high level of attenuation
  • Moisture resistant and compatible with other PPE
  • Washable and reusable
SNR: 35 dB
Sertifiseringer: EN 352-2