Face shield, Tuffmaster™
Leverandør: 3M
H8WP96
111-1635EA 1275 NOK
111-1635
Face shield, Tuffmaster™
Ansiktsskjermer
This face shield consists of headgear (H8) made of thermoplastic and clear PC visor (WP96). Both parts provide high impact resistance. Ideal for use in metalworking, manufacturing, construction, mining and transportation.
  • Adjustable ratchet suspension of the headgear allows visor to be replaced in seconds
  • Ratchet helps to provide a precise fit and comfortable head protection
  • Five position crown strap for additional support and comfort
  • Moulded PC visor provides impact and splash resistance and mounts quickly to any 3M™ headgear without tools, snaps or cams