VWR® Prague, Safety Spectacles
Leverandør: VWR Collection
111-1846EA 225 NOK
111-1846
VWR® Prague, Safety Spectacles
Vernebriller
These economical, compact cover glasses (OTG) fit over most prescription eyewear.
  • Pantoscopic adjustment allows lens angle modification
  • Adjustable temple length
Sertifiseringer: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)