VWR® Barcelona, Safety Spectacles
Leverandør: VWR Collection
111-1848EA 175 NOK
111-1848
VWR® Barcelona, Safety Spectacles
Vernebriller
Modern and stylish design glasses with sturdy polycarbonate frame.
  • Nose pad made from soft material for extra comfort and to avoid slippage
  • Soft temple dual-injected rubber to fit different head profiles
  • Pantoscopic adjustment
Sertifiseringer: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)