Safety helmets, uvex pheos B-S-WR
Leverandør: UVEX
9772.131 9772.332 9772.531 9772.031 9772.832 9772.932
111-1930EA 360 NOK
111-1930 111-1931 111-1932 111-1929 111-1933 111-1934
Safety helmets, uvex pheos B-S-WR
Hjelmer Sikkerhethjelm
Lightweight helmets made from HDPE.
  • Three variable air vents aids airflow and helps regulate the temperature within the helmet making it more comfortable to wear
  • Adapter systems for attaching uvex visors, uvex earmuffs and lamps
  • Suspension harness with rotating ratchet wheel allows the width size to be adjusted for an individual fit
  • Provides protection even in extremely low temperatures
Adjustable size range: 520 to 610 mm
Sertifiseringer: EN 397 with the additional requirement: –30 °C; MM = molten metal (splash)