Autoclavable goggles, BCAG series, BioClean Clearview™
Leverandør: Nitritex
BCAG
111-2136EA 10700 NOK
111-2136
Autoclavable goggles, BCAG series, BioClean Clearview™
Vernebriller /Goggles
BioClean Clearview™ autoclavable cleanroom goggles have a super-soft thermoplastic rubber frame to provide wearer comfort.
  • Toughened, anti-scratch, anti-fog lens
  • Super-soft frame
  • Upper vents and lower indirect ventilation system
  • Non-linting, latex-free headband
  • Can be worn over eyeglasses
  • Tested to withstand 40 autoclave cycles of 30 min duration at 121 °C/250 °F (under laboratory conditions)
  • Anti-fog performance remaining for up to 25 cycles with no degradation (under laboratory conditions)
Features super-soft thermoplastic rubber frame to provideMore Product Information