Respirator system, supplied air, S-200+
Leverandør: 3M
S200PLUS
111-2214EA 16225 NOK
111-2214
Respirator system, supplied air, S-200+
Respiratorer Vernemaskesystem
The 3M™ S-200+ supplied air respirator system is a comfortable and versatile supplied air respirator system for use with the 3M™ 6000 and 3M™ 7000 Series half and full face masks. Ideal for applications like spray painting, glass fibre fabrication, powder coating, material handling.
  • Engineered to deliver additional comfort and security the double action safety couplings help to minimise the risk of accidental disconnection during use
  • Lightweight and balanced system with a constant flow of air across the face for a cooler and more comfortable wear
  • Easy to