Supplied air regulator, Versaflo™ V-500E
Leverandør: 3M
V500E
111-2225EA 12750 NOK
111-2225
Supplied air regulator, Versaflo™ V-500E
Respiratorer Vernemaskesystem
3M™ Versaflo™ V-Series supplied air regulators offer protection against dust, mist, fumes, gases or vapours and combination hazards e.g. paint spray. Supplied air regulators do not offer protection in confined spaces, oxygen deficient atmospheres or for asbestos stripping. The 3M™ Versaflo™ V-500E supplied air regulator is a balanced, belt-worn, easy to adjust regulator specifically designed to ensure smooth airflow, safety and comfort.
  • Low pressure warning whistle
  • Integrated silencer
  • Individually adjustable, consistently controlled airflow
  • Removable,