Goggles, GoggleGear™ 500, Scotchgard™
Leverandør: 3M
GG501V
111-2327EA 565 NOK
111-2327
Goggles, GoggleGear™ 500, Scotchgard™
Vernebriller /Goggles
The vented goggles Gear 500 Scotchgard™ have a low-profile design with an adjustable elastic strap, a low profile frame and a PC lens.
  • Optical class 1, excellent UV protection
  • Longlasting Scotchgard™ Anti-fog coating prevends lens from fogging and scratches
  • Coating withstands disinfection with diluted bleach soaking or alcohol wipes without losing its anti-fog performance
For use with prescription inserts.
Sertifiseringer: EN 166:2001 ; 2C-1.2 3M 1 BT K N