Aura™ 9300+Gen3 partikkelmasker
Leverandør: 3M
9322PLUSGEN3SV 9332PLUSGEN3SV 9322PLUSGEN3 9332PLUSGEN3
111-2393EA 465 NOK
111-2393 111-2389 111-2395 111-2391
Aura™ 9300+Gen3 partikkelmasker
Respiratorer Engangsmasker Støvmasker
Aura™ particulate respirator 9300+Gen3 series is the third generation of the best-selling 3M™ Aura™ flat-fold respirator.
  • Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
  • New 3M™ Cool Flow™ Comfort valve allows a greater percentage of warm, exhaled air to escape, helping to increase comfort (compared to 3M™ Aura™ 9300+ equivalent model)
  • 3M Advanced electret (filter) media helps you breathe easier
  • Innovative tabs on the upper and lower panels simplify positioning and adjustment
  • Easy to store away with flat-fold design and