Head tops for respiratory system Versaflo™, M-206 series
Leverandør: 3M
M-206
111-2402EA 2200 NOK
111-2402
Head tops for respiratory system Versaflo™, M-206 series
Respiratorer Vernemaskesystem
Lightweight, compact and well-balanced faceshield with a comfortable faceseal for dusts, spraying and chemical processing.
  • Integrated protection from a range of respiratory, face, bumpcap, eye and hearing hazards
  • Increased control and comfort with the deflector which allows users to direct the airflow inside the headtop
  • Easy to maintain: Spare parts and accessories are quick, easy and intuitive to replace
  • Visor design combines excellent peripheral and downward vision with good optical clarity
  • Coated PC visor provides added chemical and scratch resistance