drevet luftmaskesystem (PAPR), Versaflo™ Turbo TR-300+
Leverandør: 3M
TR3600 TR302EPLUS TR315EPLUS
111-2408EA 6075 NOK
111-2408 111-2404 111-2405
drevet luftmaskesystem (PAPR), Versaflo™ Turbo TR-300+
Respiratorer Vernemaskesystem
The balanced and compact 3M™ Versaflo™ TR-300+ is an easy-to-use, versatile respirator system for particulate environments.
  • Two selectable flow rate options: 185 and 205 L/min
  • Highly efficient filter captures hazardous dust, mist and fume plus options for nuisance level gases and vapours
  • Slim, lightweight balanced design for outstanding comfort
  • Audible and LED alarm signals low battery charge or low airflow
  • Lithium ion battery with LED to indicate charge status
  • Display shows battery charge status, selected airflow and particulate filter loading status