Protective visor, anti-COVID face shield
Leverandør: MULTIROIR
26011
111-2468EA 2050 NOK
111-2468
Protective visor, anti-COVID face shield
Ansiktsskjermer
Anti-COVID personal protective visors are adjustable and comfortable, allowing for extended use. The ergonomics of the head circumference allow for convenient assembly and adjustment.
  • Facial protection from droplets, saliva, splash, oil, and dust
  • Head circumference is polypropylene material
  • Visors are made from transparent, 240 micrometer, PET (polyethylene terephthalate) material
  • Transparency guaranteed
  • Suitable for use with glasses or protective masks
  • Universal size (adjustable)
  • Delivered unmounted, easy assembly
These protective visors have beenMore Product Information