Protective screen, anti-COVID counter screen
Leverandør: MULTIROIR
L85412
111-2470EA 4375 NOK
111-2470
Protective screen, anti-COVID counter screen
Ansiktsskjermer
Anti-COVID protective screens are made from PMMA (acrylic) and are designed to provide perfect transparency.
  • PMMA material (acrylic)
  • Perfect transparency
  • High precision, laser cut
  • Round, non-cutting edges
  • 300x100 mm bottom opening
  • Protective film covering all portions of the screen
These counter screens provide additional security by preventing direct contact between the public and reception, cashier, or counter staff. Protective screens ensure that all standard exchanges can be conducted safely and completely. Screens are perfectly stable, ready toMore Product Information