Face shield, DFS1
Leverandør: Bollé Safety
PFSDFS1104
111-2494EA 0 NOK
111-2494
Face shield, DFS1
Ansiktsskjermer
The DFS1 face shield perfectly protects against splashes and droplets. Designed to fit first reponders’ needs, DFS1 is simple to use, economic, protective, and can be worn over glasses.
  • Protective band made from foam
  • Anti-fog and blue protective shield coating
  • Adapted to prescription eyewear
  • Protects against blood and bodily fluids
  • Elastic headband
Easy to assemble, the face shield is supplied as three separate components - an 0,2 mm thick PET visor with PU frame, foam band and white polyester strap.
The protective shield should be cleanedMore Product Information