Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Leverandør: 3M
MT73H7A4D10EU
111-2496EA 30800 NOK
111-2496
Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Hørselvern Hørselvern med radio
The 3M™ Peltor™ WS™ LiteCom Pro III Headset features an integrated, programmable analog/digital two-way radio, Bluetooth® Multipoint connectivity, noise-cancelling microphone, and level-dependent function for ambient listening. The headset not only helps protect against hazardous noise, but also makes it easy to communicate in noisy environments and maintain situational awareness.
  • Programmable analog/digital DMR two-way radio communication
  • Bluetooth® multipoint functionality
  • Water tight microphone (IP 68)
  • Level-dependent function for ambient listening