Safety goggles, 5X1, Vanguard PLUS
Leverandør: UNIVET
5X1.03.00.00
111-2514EA 2200 NOK
111-2514
Safety goggles, 5X1, Vanguard PLUS
Vernebriller /Goggles
Multifunctional and versatile, the X-Generation 5X1 platform ensures optimum protection and personalised use in any working condition.
  • Adjustment of temples with SoftPad technology
  • Lens with FLD (Floating Lens Design) technology
  • Patented nose pad made from anti-allergic rubber, with height adjustment, designed to reduce pressure on the nose
  • Fast interchangeable elastic band and lenses system
  • Easy assembly of the rubber gasket
Weight: 29 g
Sertifiseringer: EN 166
Bestillingsinformasjon: Wide range of lenses and corrective version are avilable.