Autoclavable cleanroom goggles, 611
Leverandør: UNIVET
611.S0.00.00.X
111-2515EA 2225 NOK
111-2515
Autoclavable cleanroom goggles, 611
Vernebriller /Goggles
These 611 cleanroom goggles are sterilisable in an autoclave with steam and also with radiation.
  • Panoramic goggles with ventilation
  • Frame and lens made of anallerginic material
  • Polycarbonate lens (2 mm)
  • Extra-wide elastic strap UNVT3PA299 is sold separately
Weight: 104 g
Sertifiseringer: EN 166