Fit test for respirators, FT10
Leverandør: 3M
FT10 FT14
111-0453EA 6925 NOK
111-0453 111-2539
Fit test for respirators, FT10
Respiratorer
Test kits and accessories designed to help your organisation meet OSHA fit testing requirements for respirators. The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapour respirator with a particulate prefilter. FT-10 uses a test solution of sodium saccharin that produces a sweet tasting aerosol.
  • Qualitative fit test kits are suitable for testing wearers of disposable respirators and half face masks fitted with particle filters
  • Kits rely on a ‘taste test’ to assess the level of fit of the respirator to the wearer’