Moulded disposable mask, FFP3, JSP LTD
Leverandør: JSP
BGZ130-000-A00
111-2586EA 450 NOK
111-2586
Moulded disposable mask, FFP3, JSP LTD
Respiratorer Engangsmasker Støvmasker
Moulded disposable respirator with comfort foam surround, adjustable nose clip, four-point adjustable harness, and exhalation valve.
  • Exhalation valve
  • Foam padded nose bridge
  • 4-point adjustable straps
  • 99% minimum filtration efficiency
A mouldable nose bridge and 360° foam area around the inside provide comfort and a secure fit. Metal-free, 4-point adjustable straps create a firm and adaptable fit.
FFP3 rating provides 99% minimum filtration efficiency.
Sertifiseringer: Conforms to EN 149.