Gas goggle, non-vented, EVO®
Leverandør: JSP
AGM030-623-000
111-2596EA 310 NOK
111-2596
Gas goggle, non-vented, EVO®
Vernebriller /Goggles
Single lens goggle for everyday protection.
  • PVC frame
  • Elastic headband
  • Anti-mist
  • Molten metal protection
  • Non vented design
  • High level of protection
Standard single lens goggle for everyday protection, engineered with anti-mist and scratch resistant single lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Goggles offer dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Sertifiseringer: EN 166; EN 170.