Goggles, uvex 9405
Leverandør: UVEX
9405.714
111-2613EA 425 NOK
111-2613
Goggles, uvex 9405
Vernebriller /Goggles
The large goggles are suitable to use with prescription eye wear and filtering half masks.
  • Soft face and nose padding ensures comfortable fit
  • Frosted upper part protects against light reflection
  • Easy headband adjustment
Sertifiseringer: EN 166, EN 170