Autoclavable Cleanroom Goggles, Isoclave IV
Leverandør: ISOFIELD IRELAND
50110
111-2618EA 20025 NOK
111-2618
Autoclavable Cleanroom Goggles, Isoclave IV
Vernebriller /Goggles
GMP Annex 1 compatible autoclavable goggle with indirect venting.
  • Autoclavable cleanroom goggle
  • ISO Class 5, GMP Annex 1 compatible
  • Anti-fog coated lens (up to 35 cycles)
  • Suitable for use over glasses
  • Non-linting strap with adjustible side clips
  • Indirect venting to prevent particle egress
The Isoclave IV google is made with a super soft thermoplastic housing to ensure comfort, and a wide view anti-fog coated lens. Suitable for autoclaving at 121 degrees Celsius for 30 minutes, the Isoclave offers up to 35 cycles on the lens and 40 on straps andMore Product Information