Heat resistant gloves, Aratherma Comfort
Leverandør: HONEYWELL SAFETY
2032681
112-0025EA 1050 NOK
112-0025
Heat resistant gloves, Aratherma Comfort
Hansker
These Kevlar® knitted gloves with cotton lining feature a green fireproof canvas cuff. Ideal for handling sharp and hot objects in a dry atmosphere.
  • Cotton lining enhances mechanical strength and absorbs perspiration
  • Highly resistant knitted construction offers protection against impact, suitable for handling heavy objects
  • Kevlar® layer offers an excellent cut resistance and thermal insulation in contact with the heat
  • Flame retardant cuff of 150 mm also protects the forearm
  • Resistance at 250 °C: 18,5 seconds; resistance at 350 °C: 13,3 seconds